0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Taurine
BP0000733438
Taurine
Bottle
APOSBUP18800-1G
New Product
:
Apollo Scientific
Taurine
APOSBUP18800-1G
:
Apollo Scientific
New Product
:
Taurine
Bottle
:
62,30 €
1 * 1 g
-
+
Pack type:
Bottle