0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
2-Phenylethanol
BP0000696569
2-Phenylethanol
Bottle
APOSBUP18776-500MG
New Product
:
Apollo Scientific
2-Phenylethanol
APOSBUP18776-500MG
:
Apollo Scientific
New Product
:
2-Phenylethanol
Bottle
:
52,10 €
1 * 500 mg
-
+
Pack type:
Bottle