Order Entry
Export
Orders LinkContactUsLinkComponent
L-Methionine ≥98%

L-Methionine ≥98%

Bottle
Catalog # APOSBUP18381-1G
New Product
Supplier:   Apollo Scientific
L-Methionine ≥98%
Catalog # APOSBUP18381-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  63-68-3
L-Methionine ≥98%

Bottle

CAS Number:  63-68-3
Price
62,30 €
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  (S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid
  • CAS Number:63-68-3
  • Formula:C₅H₁₁NO₂S
  • Molecular Weight:149,21 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications