Orders LinkContactUsLinkComponent
L-Methionine ≥98%
Bottle
  APOSBUP18381-500MG
New Product
L-Methionine ≥98%
  APOSBUP18381-500MG
New Product
 :  63-68-3
L-Methionine ≥98%
Bottle
 :  63-68-3
52,10 €
1 * 500 mg
Usage: Biochemical/life science applications
 :  (S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid
  •  :63-68-3
  •  :C₅H₁₁NO₂S
  •  :149,21 g/mol
  • Pack type:
    Bottle