Order Entry
Export
Orders LinkContactUsLinkComponent
D-​(+)​-​Phenyllactic acid ≥98%

D-​(+)​-​Phenyllactic acid ≥98%

Bottle
Catalog # APOSBUP18367-500MG
New Product
Supplier:   Apollo Scientific
D-​(+)​-​Phenyllactic acid ≥98%
Catalog # APOSBUP18367-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  7326-19-4
D-​(+)​-​Phenyllactic acid ≥98%

Bottle

CAS Number:  7326-19-4
Price
47,80 €
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  D-(+)-3-Phenyllactic acid, (R)-2-Hydroxy-3-phenylpropanoic acid, D(+)-Phenyllactic acid, D-​(+)​-​Phenyllactic acid
  • CAS Number:7326-19-4
  • Formula:C₉H₁₀O₃
  • Molecular Weight:166,17 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications