Order Entry
Export
Orders LinkContactUsLinkComponent
LM22B-10 ≥98%

LM22B-10 ≥98%

Bottle
Catalog # APOSBUP05395-25MG
New Product
Supplier:   Apollo Scientific
LM22B-10 ≥98%
Catalog # APOSBUP05395-25MG
Supplier:  Apollo Scientific
New Product
CAS Number:  342777-54-2
LM22B-10 ≥98%

Bottle

CAS Number:  342777-54-2
Price
378,00 €
Log in for availability
1 * 25 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  LM22B-10
  • CAS Number:342777-54-2
  • Formula:C₂₇H₃₃ClN₂O₄
  • Molecular Weight:485,01 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications