Order Entry
Export
Orders LinkContactUsLinkComponent
TCS-OX2-29 ≥98%

TCS-OX2-29 ≥98%

Bottle
Catalog # APOSBUP05857-100MG
New Product
Supplier:   Apollo Scientific
TCS-OX2-29 ≥98%
Catalog # APOSBUP05857-100MG
Supplier:  Apollo Scientific
New Product
CAS Number:  372523-75-6
TCS-OX2-29 ≥98%

Bottle

CAS Number:  372523-75-6
Price
633,00 €
Log in for availability
1 * 100 mg
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  (2S)-1-(3,4-Dihydro-6,7-dimethoxy-2(1H)-isoquinolinyl)-3,3-dimethyl-2-[(4-pyridinylmethyl)amino]-1-butanone hydrochloride
  • CAS Number:372523-75-6
  • Formula:C₂₃H₃₁N₃O₃
  • Molecular Weight:397,51 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications