Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
TCS OX2 29
BP0000691836
Print
TCS-OX2-29
Bottle
Catalog # APOSBUP05857-25MG
New Product
Supplier:
Apollo Scientific
TCS-OX2-29
Catalog # APOSBUP05857-25MG
Supplier:
Apollo Scientific
New Product
CAS Number:
Return to Top
TCS-OX2-29
Bottle
Specifications
Jump to:
Specifications
CAS Number:
Price
241,00 €
Log in
for availability
1 * 25 mg
-
+
Other Product Family Options Available
Return Policy
Product Details & Documents
Specifications
Specifications
Pack type:
Bottle
Cat. No.:
APOSBUP05857-25MG
Specifications