Order Entry
Export
Orders LinkContactUsLinkComponent
Degrasyn ≥98%

Degrasyn ≥98%

Bottle
Catalog # APOSBUP06025-50MG
New Product
Supplier:   Apollo Scientific
Degrasyn ≥98%
Catalog # APOSBUP06025-50MG
Supplier:  Apollo Scientific
New Product
CAS Number:  856243-80-6
Degrasyn ≥98%

Bottle

CAS Number:  856243-80-6
Price
479,00 €
Log in for availability
1 * 50 mg
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  Degrasyn, (S,E)-3-(6-Bromopyridin-2-yl)-2-cyano-N-(1-phenylbutyl)acrylamide
  • CAS Number:856243-80-6
  • Formula:C₁₉H₁₈BrN₃O
  • Molecular Weight:384,27 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications