Order Entry
Export
Orders LinkContactUsLinkComponent
 

4-Ethylcatechol ≥98%

Bottle
Catalog # APOSBUP06059-100MG
New Product
Supplier:   Apollo Scientific
4-Ethylcatechol ≥98%
Catalog # APOSBUP06059-100MG
Supplier:  Apollo Scientific
New Product
CAS Number:  1124-39-6
4-Ethylcatechol ≥98%

Bottle

CAS Number:  1124-39-6
Price
378,00 €
Log in for availability
1 * 100 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  4-Ethylbenzene-1,2-diol, 4-Ethylcatechol
  • CAS Number:1124-39-6
  • Formula:C₈H₁₀O₂
  • Molecular Weight:138,16 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications