4-Ethylcatechol ≥98%
Bottle
Catalog # APOSBUP06059-50MG
New Product
Supplier: Apollo Scientific
4-Ethylcatechol ≥98%
Catalog # APOSBUP06059-50MG
Supplier: Apollo Scientific
New Product
CAS Number: 1124-39-6
Jump to:
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyms: 4-Ethylbenzene-1,2-diol, 4-Ethylcatechol
- CAS Number:1124-39-6
- Formula:C₈H₁₀O₂
- Molecular Weight:138,16 g/mol
Specifications
Specifications
- Pack type:Bottle



