Order Entry
Export
Orders LinkContactUsLinkComponent
 

A-196 ≥98%

Bottle
Catalog # APOSBUP05604-25MG
New Product
Supplier:   Apollo Scientific
A-196 ≥98%
Catalog # APOSBUP05604-25MG
Supplier:  Apollo Scientific
New Product
CAS Number:  1982372-88-2
A-196 ≥98%

Bottle

CAS Number:  1982372-88-2
Price
251,00 €
Log in for availability
1 * 25 mg
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  A-196
  • CAS Number:1982372-88-2
  • Formula:C₁₈H₁₆Cl₂N₄
  • Molecular Weight:359,25 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications