Order Entry
Export
Orders LinkContactUsLinkComponent
m-Coumaric acid ≥98%

m-Coumaric acid ≥98%

Bottle
Catalog # APOSBUP18246-500MG
New Product
Supplier:   Apollo Scientific
m-Coumaric acid ≥98%
Catalog # APOSBUP18246-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  588-30-7
m-Coumaric acid ≥98%

Bottle

CAS Number:  588-30-7
Price
63,70 €
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  m-Hydroxycinnamic acid, 3-(3-Hydroxyphenyl)acrylic acid, m-Coumaric acid
  • CAS Number:588-30-7
  • Formula:C₉H₈O₃
  • Molecular Weight:164,16 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications