Order Entry
Export
Orders LinkContactUsLinkComponent
 

Phenolphthalein ≥98%

Bottle
Catalog # APOSBUP18279-500MG
New Product
Supplier:   Apollo Scientific
Phenolphthalein ≥98%
Catalog # APOSBUP18279-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  77-09-8
Phenolphthalein ≥98%

Bottle

CAS Number:  77-09-8
Price
53,60 €
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
  • CAS Number:77-09-8
  • Formula:C₂₀H₁₄O₄
  • Molecular Weight:318,32 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications