Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
SC-43
BP0000698189
Print
SC-43
Bottle
Catalog # APOSBUP04224-25MG
New Product
Supplier:
Apollo Scientific
SC-43
Catalog # APOSBUP04224-25MG
Supplier:
Apollo Scientific
New Product
CAS Number:
Return to Top
SC-43
Bottle
Specifications
Jump to:
Specifications
CAS Number:
Price
684,00 €
Log in
for availability
1 * 25 mg
-
+
Other Product Family Options Available
Return Policy
Product Details & Documents
Specifications
Specifications
Pack type:
Bottle
Specifications