5-hydroxytryptophan ≥98%
Bottle
Catalog # APOSBUP00053-1G
New Product
Supplier: Apollo Scientific
5-hydroxytryptophan ≥98%
Catalog # APOSBUP00053-1G
Supplier: Apollo Scientific
New Product
CAS Number: 56-69-9
Jump to:
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyms: DL-5-Hydroxytryptophan, 2-Amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid
- CAS Number:56-69-9
- Formula:C₁₁H₁₂N₂O₃
- Molecular Weight:220,22 g/mol
Specifications
Specifications
- Pack type:Bottle



