Order Entry
Export
Orders LinkContactUsLinkComponent
 

5-hydroxytryptophan ≥98%

Bottle
Catalog # APOSBUP00053-20MG
New Product
Supplier:   Apollo Scientific
5-hydroxytryptophan ≥98%
Catalog # APOSBUP00053-20MG
Supplier:  Apollo Scientific
New Product
CAS Number:  56-69-9
5-hydroxytryptophan ≥98%

Bottle

CAS Number:  56-69-9
Price
63,70 €
Log in for availability
1 * 20 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  DL-5-Hydroxytryptophan, 2-Amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid
  • CAS Number:56-69-9
  • Formula:C₁₁H₁₂N₂O₃
  • Molecular Weight:220,22 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications