Order Entry
Export
Orders LinkContactUsLinkComponent
NBD-556 ≥98%

NBD-556 ≥98%

Bottle
Catalog # APOSBUP10127-100MG
New Product
Supplier:   Apollo Scientific
NBD-556 ≥98%
Catalog # APOSBUP10127-100MG
Supplier:  Apollo Scientific
New Product
CAS Number:  333353-44-9
NBD-556 ≥98%

Bottle

CAS Number:  333353-44-9
Price
274,00 €
Log in for availability
1 * 100 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  N1-(4-Chlorophenyl)-N2-(2,2,6,6-tetramethylpiperidin-4-yl)oxalamide, N1-(4-Chlorophenyl)-N2-(2,2,6,6-tetramethyl-4-piperidinyl)ethanediamide
  • CAS Number:333353-44-9
  • Formula:C₁₇H₂₄ClN₃O₂
  • Molecular Weight:337,84 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications