NBD-556 ≥98%
Bottle
Catalog # APOSBUP10127-25MG
New Product
Supplier: Apollo Scientific
NBD-556 ≥98%
Catalog # APOSBUP10127-25MG
Supplier: Apollo Scientific
New Product
CAS Number: 333353-44-9
Jump to:
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyms: N1-(4-Chlorophenyl)-N2-(2,2,6,6-tetramethylpiperidin-4-yl)oxalamide, N1-(4-Chlorophenyl)-N2-(2,2,6,6-tetramethyl-4-piperidinyl)ethanediamide
- CAS Number:333353-44-9
- Formula:C₁₇H₂₄ClN₃O₂
- Molecular Weight:337,84 g/mol
Specifications
Specifications
- Pack type:Bottle



