O4I1 ≥95%
Bottle
Catalog # APOSBUP11026-200MG
New Product
Supplier: Apollo Scientific
O4I1 ≥95%
Catalog # APOSBUP11026-200MG
Supplier: Apollo Scientific
New Product
CAS Number: 175135-47-4
Jump to:
Product Details & Documents
Usage: Biochemical/life science applications.
Synonyms: 4-[(4-Methoxyphenyl)methoxy]benzeneacetonitrile, 2-(4-((4-Methoxybenzyl)oxy)phenyl)acetonitrile, O4I1
- CAS Number:175135-47-4
- Formula:C₁₆H₁₅NO₂
- Molecular Weight:253,3 g/mol
Specifications
Specifications
- Pack type:Bottle



