Order Entry
Export
Orders LinkContactUsLinkComponent
O4I1 ≥95%

O4I1 ≥95%

Bottle
Catalog # APOSBUP11026-50MG
New Product
Supplier:   Apollo Scientific
O4I1 ≥95%
Catalog # APOSBUP11026-50MG
Supplier:  Apollo Scientific
New Product
CAS Number:  175135-47-4
O4I1 ≥95%

Bottle

CAS Number:  175135-47-4
Price
102,00 €
Log in for availability
1 * 50 mg
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  4-[(4-Methoxyphenyl)methoxy]benzeneacetonitrile, 2-(4-((4-Methoxybenzyl)oxy)phenyl)acetonitrile, O4I1
  • CAS Number:175135-47-4
  • Formula:C₁₆H₁₅NO₂
  • Molecular Weight:253,3 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications