Order Entry
Export
Orders LinkContactUsLinkComponent
Antitubercular agent-41 ≥98%

Antitubercular agent-41 ≥98%

Bottle
Catalog # APOSBUP04667-100MG
New Product
Supplier:   Apollo Scientific
Antitubercular agent-41 ≥98%
Catalog # APOSBUP04667-100MG
Supplier:  Apollo Scientific
New Product
CAS Number:  901032-23-3
Antitubercular agent-41 ≥98%

Bottle

CAS Number:  901032-23-3
Price
856,00 €
Log in for availability
1 * 100 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  [4-(4-fluorophenyl)-1-piperazinyl](6-methoxyfuro[2,3-b]quinolin-2-yl)-methanone
  • CAS Number:901032-23-3
  • Formula:C₂₃H₂₀FN₃O₃
  • Molecular Weight:405,42 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications