Antitubercular agent-41 ≥98%
Bottle
Catalog # APOSBUP04667-25MG
New Product
Supplier: Apollo Scientific
Antitubercular agent-41 ≥98%
Catalog # APOSBUP04667-25MG
Supplier: Apollo Scientific
New Product
CAS Number: 901032-23-3
Jump to:
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyms: [4-(4-fluorophenyl)-1-piperazinyl](6-methoxyfuro[2,3-b]quinolin-2-yl)-methanone
- CAS Number:901032-23-3
- Formula:C₂₃H₂₀FN₃O₃
- Molecular Weight:405,42 g/mol
Specifications
Specifications
- Pack type:Bottle



