Order Entry
Export
Orders LinkContactUsLinkComponent
 

L-Thyroxine ≥95%

Bottle
Catalog # APOSBUP17744-500MG
New Product
Supplier:   Apollo Scientific
L-Thyroxine ≥95%
Catalog # APOSBUP17744-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  51-48-9
L-Thyroxine ≥95%

Bottle

CAS Number:  51-48-9
Price
68,10 €
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  (S)-2-Amino-3-(4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl)propanoic acid
  • CAS Number:51-48-9
  • Formula:C₁₅H₁₁I₄NO₄
  • Molecular Weight:776,87 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications