L-Thyroxine ≥95%
Bottle
Catalog # APOSBUP17744-200MG
New Product
Supplier: Apollo Scientific
L-Thyroxine ≥95%
Catalog # APOSBUP17744-200MG
Supplier: Apollo Scientific
New Product
CAS Number: 51-48-9
Jump to:
Product Details & Documents
Usage: Biochemical/life science applications.
Synonyms: (S)-2-Amino-3-(4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl)propanoic acid
- CAS Number:51-48-9
- Formula:C₁₅H₁₁I₄NO₄
- Molecular Weight:776,87 g/mol
Specifications
Specifications
- Pack type:Bottle



