Order Entry
Export
Orders LinkContactUsLinkComponent
 

Adenosine ≥98%

Bottle
Catalog # APOSBUP18498-500MG
New Product
Supplier:   Apollo Scientific
Adenosine ≥98%
Catalog # APOSBUP18498-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  58-61-7
Adenosine ≥98%

Bottle

CAS Number:  58-61-7
Price
59,40 €
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside
  • CAS Number:58-61-7
  • Formula:C₁₀H₁₃N₅O₄
  • Molecular Weight:267,24 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications