Order Entry
Export
Orders LinkContactUsLinkComponent
 

Dexamethasone acetate ≥98%

Bottle
Catalog # APOSBUP18470-1G
New Product
Supplier:   Apollo Scientific
Dexamethasone acetate ≥98%
Catalog # APOSBUP18470-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  1177-87-3
Dexamethasone acetate ≥98%

Bottle

CAS Number:  1177-87-3
Price
98,40 €
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  11β,17α-Dihydroxy-21-(ethanoyloxy)-9-fluor-16α-methyl-1,4-pregnadien-3,20-dion, Dexamethasone acetate
  • CAS Number:1177-87-3
  • Formula:C₂₄H₃₁FO₆
  • Molecular Weight:434,5 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications