Order Entry
Export
Orders LinkContactUsLinkComponent
1-Hexanol ≥98%

1-Hexanol ≥98%

Bottle
Catalog # APOSBUP19339-10G
New Product
Supplier:   Apollo Scientific
1-Hexanol ≥98%
Catalog # APOSBUP19339-10G
Supplier:  Apollo Scientific
New Product
CAS Number:  111-27-3
1-Hexanol ≥98%

Bottle

CAS Number:  111-27-3
Price
137,00 €
Log in for availability
1 * 10 g
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  Hexyl alcohol, Pentyl carbinol, 1-Hexyl alcohol, 1-Hydroxy hexane
  • CAS Number:111-27-3
  • Formula:C₆H₁₄O
  • Molecular Weight:102,17 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications