0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
PYR-41
BP0000725120
PYR-41
Bottle
APOSBUP15362-100MG
New Product
:
Apollo Scientific
PYR-41
APOSBUP15362-100MG
:
Apollo Scientific
New Product
:
PYR-41
Bottle
:
633,00 €
1 * 100 mg
-
+
Pk:
Bottle