Order Entry
Export
Orders LinkContactUsLinkComponent
Cinnamaldehyde ≥80%

Cinnamaldehyde ≥80%

Bottle
Catalog # APOSBUP17096-5G
Irritant
New Product
Supplier:   Apollo Scientific
Cinnamaldehyde ≥80%
Catalog # APOSBUP17096-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  104-55-2
Cinnamaldehyde ≥80%

Bottle

CAS Number:  104-55-2
Price
261,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  trans-Cinnamaldehyde, 3-Phenylacrylaldehyde
  • CAS Number:104-55-2
  • Formula:C₉H₈O
  • MDL Number:MFCD00007000
  • Molecular Weight:132,16 g/mol
  • EINECS:203-213-9
Specifications

Specifications

  • Pack type:
    Bottle

Specifications