Order Entry
Export
Orders LinkContactUsLinkComponent
Squalene ≥98%

Squalene ≥98%

Bottle
Catalog # APOSBUP17811-500MG
New Product
Supplier:   Apollo Scientific
Squalene ≥98%
Catalog # APOSBUP17811-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  111-02-4
Squalene ≥98%

Bottle

CAS Number:  111-02-4
Price
280,00 €
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  2,6,10,15,19,23-Hexamethyltetracosa-2,6,10,14,18,22-hexaene
  • CAS Number:111-02-4
  • Formula:C₃₀H₅₀
  • Molecular Weight:410,72 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications