0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
2,2-Dimethyl-4-oxo-3,7,10,13,16-pentaoxanonadecan-19-oic acid
BP0000737120
acid-PEG4-C2-BOC
acid-PEG4-C2-BOC
Bottle
APOSOR1026354-5G
New Product
:
Apollo Scientific
acid-PEG4-C2-BOC
APOSOR1026354-5G
:
Apollo Scientific
New Product
:
Expand image
acid-PEG4-C2-BOC
Bottle
Add to cart
:
1 360,00 €
Unit of Measure
1 * 5 g
-
+
Add to cart
Pack type:
Bottle