Order Entry
Export
Orders LinkContactUsLinkComponent
acid-PEG4-C2-BOC 98%

acid-PEG4-C2-BOC 98%

Bottle
Catalog # APOSOR1026354-1G
New Product
Supplier:   Apollo Scientific
acid-PEG4-C2-BOC 98%
Catalog # APOSOR1026354-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  1835759-85-7
acid-PEG4-C2-BOC 98%

Bottle

CAS Number:  1835759-85-7
Price
368,00 €
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  Acid-PEG4-C2-Boc
  • CAS Number:1835759-85-7
  • Formula:C₁₆H₃₀O₈
  • Molecular Weight:350,4 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications