Order Entry
Export
Orders LinkContactUsLinkComponent
Protocatechualdehyde ≥98%

Protocatechualdehyde ≥98%

Bottle
Catalog # APOSBUP17058-10G
New Product
Supplier:   Apollo Scientific
Protocatechualdehyde ≥98%
Catalog # APOSBUP17058-10G
Supplier:  Apollo Scientific
New Product
CAS Number:  139-85-5
Protocatechualdehyde ≥98%

Bottle

CAS Number:  139-85-5
Price
563,00 €
Log in for availability
1 * 10 g
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  Protocatechuic aldehyde, Protocatechualdehyde
  • CAS Number:139-85-5
  • Formula:C₇H₆O₃
  • Molecular Weight:138,12 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications