0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
D(+)-Mannose
D-Mannose ≥95%
D-Mannose ≥95%
Bottle
APOSBUP16831-500MG
New Product
:
Apollo Scientific
D-Mannose ≥95%
APOSBUP16831-500MG
:
Apollo Scientific
New Product
:
3458-28-4
D-Mannose ≥95%
Bottle
:
3458-28-4
116,00 €
1 * 500 mg
-
+
Usage: Biochemical/life science applications
:
(2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
:
3458-28-4
:
C₆H₁₂O₆
:
180,16 g/mol
Pack type:
Bottle