Order Entry
Export
Orders LinkContactUsLinkComponent
D-Mannose ≥95%

D-Mannose ≥95%

Bottle
Catalog # APOSBUP16831-100MG
New Product
Supplier:   Apollo Scientific
D-Mannose ≥95%
Catalog # APOSBUP16831-100MG
Supplier:  Apollo Scientific
New Product
CAS Number:  3458-28-4
D-Mannose ≥95%

Bottle

CAS Number:  3458-28-4
Price
50,70 €
Log in for availability
1 * 100 mg
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
  • CAS Number:3458-28-4
  • Formula:C₆H₁₂O₆
  • Molecular Weight:180,16 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications