Order Entry
Export
Orders LinkContactUsLinkComponent
PD98059 ≥98%

PD98059 ≥98%

Bottle
Catalog # APOSBUP15496-200MG
New Product
Supplier:   Apollo Scientific
PD98059 ≥98%
Catalog # APOSBUP15496-200MG
Supplier:  Apollo Scientific
New Product
CAS Number:  167869-21-8
PD98059 ≥98%

Bottle

CAS Number:  167869-21-8
Price
355,00 €
Log in for availability
1 * 200 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  PD98059, 2'-Amino-3'-methoxyflavone, 2-(2-Amino-3-methoxyphenyl)chromone, PD 98059, 2-(2-Amino-3-methoxyphenyl)-4H-1-benzopyran-4-one, 2-(2-Amino-3-methoxyphenyl)-4H-chromen-4-one
  • CAS Number:167869-21-8
  • Formula:C₁₆H₁₃NO₃
  • Molecular Weight:267,28 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications