Order Entry
Export
Orders LinkContactUsLinkComponent
 

Sodium Acetate 98% buffer solution

Bottle
Catalog # APOSOR1027993-100G
New Product
Supplier:   Apollo Scientific
Sodium Acetate 98% buffer solution
Catalog # APOSOR1027993-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  126-96-5
Sodium Acetate 98% buffer solution

Bottle

CAS Number:  126-96-5
Price
24,60 €
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  Acetic acid sodium salt
  • CAS Number:126-96-5
  • Formula:C₄H₇NaO₄
  • Molecular Weight:142,09 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications