Sodium Acetate 98% buffer solution
Bottle
Catalog # APOSOR1027993-25G
New Product
Supplier: Apollo Scientific
Sodium Acetate 98% buffer solution
Catalog # APOSOR1027993-25G
Supplier: Apollo Scientific
New Product
CAS Number: 126-96-5
Jump to:
Product Details & Documents
Usage: Organic building block/ reagent.
Synonyms: Acetic acid sodium salt
- CAS Number:126-96-5
- Formula:C₄H₇NaO₄
- Molecular Weight:142,09 g/mol
Specifications
Specifications
- Pack type:Bottle



