0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Benzalkonium chloride
BP0000739086
Benzalkonium chloride
Bottle
APOSOR1029921-10KG
New Product
:
Apollo Scientific
Benzalkonium chloride
APOSOR1029921-10KG
:
Apollo Scientific
New Product
:
Benzalkonium chloride
Bottle
:
2 150,00 €
1 * 10 kg
-
+
Pack type:
Bottle