Order Entry
Export
Orders LinkContactUsLinkComponent
Benzalkonium chloride 80%

Benzalkonium chloride 80%

Bottle
Catalog # APOSOR1029921-100G
Corrosive
Environment
Irritant
New Product
Supplier:   Apollo Scientific
Benzalkonium chloride 80%
Catalog # APOSOR1029921-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  68424-85-1
Benzalkonium chloride 80%

Bottle

CAS Number:  68424-85-1
Price
91,20 €
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  Quaternary ammonium compounds, benzyl-C8-18-alkyldimethyl, chlorides, Alkylbenzyldimethylammonium chloride
  • CAS Number:68424-85-1
  • MDL Number:MFCD00145757
  • EINECS:270-325-2
  • UN:1760
Specifications

Specifications

  • Pack type:
    Bottle

Specifications