0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Benzalkonium chloride
BP0000739086
Benzalkonium chloride
Benzalkonium chloride
Bottle
APOSOR1029921-5G
New Product
:
Apollo Scientific
Benzalkonium chloride
APOSOR1029921-5G
:
Apollo Scientific
New Product
:
Expand image
Benzalkonium chloride
Bottle
Add to cart
:
21,80 €
Unit of Measure
1 * 5 g
-
+
Add to cart
Pack type:
Bottle