Order Entry
Export
Orders LinkContactUsLinkComponent
TRAM 39 ≥98%

TRAM 39 ≥98%

Bottle
Catalog # APOSBUP03170-25MG
New Product
Supplier:   Apollo Scientific
TRAM 39 ≥98%
Catalog # APOSBUP03170-25MG
Supplier:  Apollo Scientific
New Product
CAS Number:  197525-99-8
TRAM 39 ≥98%

Bottle

CAS Number:  197525-99-8
Price
283,00 €
Log in for availability
1 * 25 mg
Product Details & Documents

Usage: Biochemical/life science applications.

  • CAS Number:197525-99-8
  • Formula:C₂₀H₁₄ClN
  • Molecular Weight:303,78 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications