0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
N,N'-(9-((4-(Dimethylamino)phenyl)amino)acridine-3,6-diyl)bis(3-(pyrrolidin-1-yl)propanamide)
BP0000728216
Braco-19
Braco-19
APOSBUP03340-100MG
New Product
:
Apollo Scientific
Braco-19
APOSBUP03340-100MG
:
Apollo Scientific
New Product
:
Expand image
Braco-19
Add to cart
:
1 200,00 €
Unit of Measure
1 * 100 mg
-
+
Add to cart