Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
N,N'-(9-((4-(Dimethylamino)phenyl)amino)acridine-3,6-diyl)bis(3-(pyrrolidin-1-yl)propanamide)
BP0000728216
Braco-19
Print
Braco-19
Catalog # APOSBUP03340-25MG
New Product
Supplier:
Apollo Scientific
Braco-19
Catalog # APOSBUP03340-25MG
Supplier:
Apollo Scientific
New Product
CAS Number:
Expand image
Return to Top
Braco-19
Add to cart
CAS Number:
Price
526,00 €
Log in
for availability
Unit of Measure
1 * 25 mg
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents