0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
1,1'-((Butane-1,4-diylbis(4,1-phenylene))bis(methylene))bis(4-(dimethylamino)pyridin-1-ium) bromide
BP0000687695
MN58b
Bottle
APOSBUP08618-25MG
New Product
:
Apollo Scientific
MN58b
APOSBUP08618-25MG
:
Apollo Scientific
New Product
:
MN58b
Bottle
:
1 970,00 €
1 * 25 mg
-
+
Pack type:
Bottle