0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
2-(2-Chlorophenyl)-2,2-diphenylacetonitrile
BP0000691671
TRAM 39
Bottle
APOSBUP03170-100MG
New Product
:
Apollo Scientific
TRAM 39
APOSBUP03170-100MG
:
Apollo Scientific
New Product
:
TRAM 39
Bottle
:
710,00 €
1 * 100 mg
-
+
Pack type:
Bottle