0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
STF-31
BP0000683655
STF-31
Bottle
APOSBUP07777-100MG
New Product
:
Apollo Scientific
STF-31
APOSBUP07777-100MG
:
Apollo Scientific
New Product
:
STF-31
Bottle
:
856,00 €
1 * 100 mg
-
+
Pack type:
Bottle