0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
(±)-α-Lipoic acid
BP0000728040
DL-alpha-Lipoic acid
APOSBID9805-10KG
New Product
:
Apollo Scientific
DL-alpha-Lipoic acid
APOSBID9805-10KG
:
Apollo Scientific
New Product
:
DL-alpha-Lipoic acid
:
1 770,00 €
1 * 10 kg
-
+