Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
(±)-α-Lipoic acid
BP0000728040
Print
DL-alpha-Lipoic acid
Catalog # APOSBID9805-100G
New Product
Supplier:
Apollo Scientific
DL-alpha-Lipoic acid
Catalog # APOSBID9805-100G
Supplier:
Apollo Scientific
New Product
CAS Number:
Return to Top
DL-alpha-Lipoic acid
CAS Number:
Price
60,80 €
Log in
for availability
1 * 100 g
-
+
Other Product Family Options Available
Return Policy
Product Details & Documents